Anti-ARIH2 Rabbit Polyclonal Antibody

Supplier: Proteintech
15006-1-AP-150UL
10083-112EA 402 USD
10083-112
Anti-ARIH2 Rabbit Polyclonal Antibody
Antibodies
ARIH2(Protein ariadne-2 homolog) is also named as ARI2, TRIAD1 and belongs to the RBR family and ariadne subfamily .It mediates 'Lys-48'-and 'Lys-63'-linked polyubiquitination and subsequent proteasomal degradation of modified proteins.

Western Blot: HEK-293 Cells, 1:1000-1:10000

Type: Primary
Antigen: ARIH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR