Anti-PRSS2 Rabbit Polyclonal Antibody

Supplier: Proteintech
15005-1-AP-150UL
10092-854EA 402 USD
10092-854
Anti-PRSS2 Rabbit Polyclonal Antibody
Antibodies
The human pancreas produces three isoforms of trypsinogen encoded by separate genes, the PRSS1 (protease, serine 1), PRSS2, and PRSS3 genes. PRSS2 is also named as TRY2(trypsin-2), TRYP2. In the ileum, it may be involved in defensin processing, including DEFA5. The preproprotein comprises 247 amino acids, including a 15-amino acid signal peptide and an 8-amino acid activation peptide.

Western Blot: Raji Cells, 1:500-1:5000

Type: Primary
Antigen: PRSS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR