Anti-HIRIP3 Rabbit Polyclonal Antibody

Supplier: Proteintech
14992-1-AP-150UL
10088-262EA 402 USD
10088-262
Anti-HIRIP3 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Skeletal Muscle Tissue, 1:200-1:2000; IHC: Human ovary Tissue, 1:20-1:200

Type: Primary
Antigen: HIRIP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR