Anti-PPA1 Rabbit Polyclonal Antibody

Supplier: Proteintech
14985-1-AP-150UL
10092-616EA 402 USD
10092-616
Anti-PPA1 Rabbit Polyclonal Antibody
Antibodies
PPA1(Inorganic pyrophosphatase) is also named as IOPPP, PP and PPase. It maintains the thermodynamic favorability of these reactions by catalyzing the hydrolysis of pyrophosphates into organic phosphates, which are then exported across the cell membrane.The erythrocyte inorganic pyrophosphatase molecule would appear to be a dimer.

Western Blot: Human Heart Tissue, 1:500-1:5000; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: PPA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR