Anti-GAL3 Rabbit Polyclonal Antibody

Supplier: Proteintech
14979-1-AP-150UL
10087-338EA 537.7 USD
10087-338
Anti-GAL3 Rabbit Polyclonal Antibody
Antibodies
Galectins are a family of animal lectins defined by shared characteristic amino-acid sequences and affinity for β-galactose-containing oligosac-charides . Galectin-3 contains one carbohydrate recognition domain (CRD) and a proline- and glycine-rich N-terminal domain through which is able to form oligomers. Galectin-3 is widely expressed in many normal tissues and a variety of tumors. It is found intracellularly in nucleus and cytoplasm or secreted outside of cell, being present on the cell surface or in the extracellular space . Galectin-3 is involved in various biological processes including cell growth, adhesion, differentiation, apoptosis, angiogenesis, immune response, neoplastic transformation and metastasis .

Western Blot:NIH/3T3 Cells, 1:200-1:2000; IHC: Human thyroid cancer Tissue, 1:20-1:200; IP: MCF-7 Cells, 1:200-1:2000

Type: Primary
Antigen: Galectin-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR