Anti-SNRPE Rabbit Polyclonal Antibody

Supplier: Proteintech
14977-1-AP-150UL
10094-956EA 402 USD
10094-956
Anti-SNRPE Rabbit Polyclonal Antibody
Antibodies
The 4 major small nuclear ribonucleoprotein particles (snRNPs), U1, U2, U4/U6, and U5, are essential components of the eukaryotic pre-mRNA splicing machinery. These snRNPs share 8 proteins which form the snRNP structural core: B/B-prime (SNRPB), D1 (SNRPD1), D2 (SNRPD2), D3 (SNRPD3), E (SNRPE), F (SNRPF), and G (SNRPG). These proteins have a role in the biogenesis of the snRNPs.

Western Blot: Human Heart Tissue, 1:500-1:5000; IHC: Human breast cancer Tissue, 1:20-1:200; FC: MCF-7 Cells, N/A

Type: Primary
Antigen: SNRPE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR