Anti-UQCRQ Rabbit Polyclonal Antibody

Supplier: Proteintech
14975-1-AP-150UL
10096-592EA 402 USD
10096-592
Anti-UQCRQ Rabbit Polyclonal Antibody
Antibodies
UQCRQ(Cytochrome b-c1 complex subunit 8) belongs to the UQCRQ/QCR8 family. UQCRQ is a ubiquinone-binding protein localized to the cytochrome bc1 region of the mitochondrial respiratory chain. The deduced 110-amino acid protein has a relatively hydrophobic and basic N-terminal region, an aspartic acid-rich middle region, and a glutamic acid- and lysine-rich C-terminal region.

Western Blot: Human Liver Tissue, 1:500-1:5000; IHC: Human Liver cancer Tissue, 1:20-1:200

Type: Primary
Antigen: UQCRQ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR