Anti-DDAH2 Rabbit Polyclonal Antibody

Supplier: Proteintech
14966-1-AP-150UL
10085-730EA 402 USD
10085-730
Anti-DDAH2 Rabbit Polyclonal Antibody
Antibodies
The DDAH family of proteins is formed by two isoforms, DDAH-1 and DDAH-2, both of which utilize ADMA as their substrate and increase NOS activity. DDAH-2,also named as DDAH, G6A, NG30, is the most prevalent isoform in blood vessels and endothelium and is important for embryonic development. It has also been linked to other diseases, such as hypertension, cerebral hemorrhage, cardiac disease, and diabetes. In cultured human endothelial cells,DDAH-2 had been found only in the cytosol, but in the nuclei of rat vascular smooth muscle cells..

Western Blot: Human Heart Tissue, 1:200-1:2000; IHC: Human Colon cancer Tissue, 1:20-1:200

Type: Primary
Antigen: DDAH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR