Anti-RPS15 Rabbit Polyclonal Antibody

Supplier: Proteintech
14957-1-AP-150UL
10093-944EA 402 USD
10093-944
Anti-RPS15 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Liver Tissue, 1:500-1:5000; IHC: Human Liver cancer Tissue, 1:20-1:200; IF: Hela Cells, 1:20-1:200

Type: Primary
Antigen: RPS15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR