Anti-Kir6.1 Rabbit Polyclonal Antibody

Supplier: Proteintech
14954-1-AP-150UL
10089-278EA 402 USD
10089-278
Anti-Kir6.1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse spleen Tissue, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: Kir6.1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR