Anti-RPS27A Rabbit Polyclonal Antibody

Supplier: Proteintech
14946-1-AP-150UL
10093-960EA 402 USD
10093-960
Anti-RPS27A Rabbit Polyclonal Antibody
Antibodies
RPS27A, also named as UBA80 and UBCEP1, can be cleaved into two chains Ubiquitin and 40S ribosomal protein S27a. Ubiquitin is a highly conserved 76-amino acid protein that plays a key role in protein degradation. Ubiquitin is synthesized as precursor proteins that consist either of polyubiquitin chains that are cleaved into moieties of the UBB or UBC types, or single ubiquitin moieties fused 5-prime to unrelated carboxyl extension proteins (UBA type) such as UBA52 and UBA80 (RPS27A)

Western Blot: Mouse cerebellum Tissue, 1:200-1:1000; IHC: Human endometrial cancer Tissue, 1:20-1:200

Type: Primary
Antigen: RPS27A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR