Anti-BLMH Rabbit Polyclonal Antibody

Supplier: Proteintech
14941-1-AP-150UL
10083-626EA 402 USD
10083-626
Anti-BLMH Rabbit Polyclonal Antibody
Antibodies
BLMH(Bleomycin hydrolase) is also named as BH, BMH and belongs to the peptidase C1 family. The gene is expressed in many human tissues with elevated expression levels found in testis, skeletal muscle, and pancreas, and low levels of expression in colon and peripheral blood leukocytes. The protein catalyzes the inactivation of the antitumor drug BLM (a glycopeptide) by hydrolyzing the carboxamide bond of its B-aminoalaninamide moiety thus protecting normal and malignant cells from BLM toxicity. BH-immunoreactivity is essentially located in the cytoplasm and at the outer membrane of the rough endoplasmic reticulum of epithelial cells of the kidney, as well as in that of hepatocytes of the liver.

Western Blot: Human Colon Tissue, 1:500-1:5000; IHC: Human Colon cancer Tissue, 1:20-1:200; IF: Hela Cells, 1:20-1:200

Type: Primary
Antigen: BLMH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR