Anti-GNPAT Rabbit Polyclonal Antibody

Supplier: Proteintech
14931-1-AP-150UL
10087-714EA 402 USD
10087-714
Anti-GNPAT Rabbit Polyclonal Antibody
Antibodies
GNPAT, also named as DAPAT and DHAPAT, belongs to the GPAT/DAPAT family. It is a key enzyme in the biosynthesis of ether phospholipids. GNPAT is localized exclusively within peroxisomes. Full GNPAT activity depends not only on the presence of AGPS, but also on the integrity of substrate channeling from GNPAT to AGPS. This antibody recognize the 69kd GNPAT protein.

Western Blot: HepG2 Cells, 1:200-1:2000; IF: MCF-7 Cells, 1:20-1:200

Type: Primary
Antigen: GNPAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR