Anti-FAH Rabbit Polyclonal Antibody

Supplier: Proteintech
14928-1-AP-150UL
10086-770EA 402 USD
10086-770
Anti-FAH Rabbit Polyclonal Antibody
Antibodies
FAH is also named as FAA(fumarylacetoacetase), the last enzyme in tyrosine degradation, functionally important in the liver and kidney. FAH is a metabolic enzyme catalyzing the last step of tyrosine and phenylalanine catabolism: the hydrolysis of fumarylacetoacetate into acetoacetate and fumarate. In humans, deficiency of this activity is associated with the metabolic disease hereditary tyrosinaemia type I, which is also known as hepatorenal tyrosinaemia.

Western Blot: Mouse Liver Tissue, 1:200-1:2000

Type: Primary
Antigen: FAH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR