Anti-ATP6V1D Rabbit Polyclonal Antibody

Supplier: Proteintech
14920-1-AP-150UL
10083-370EA 402 USD
10083-370
Anti-ATP6V1D Rabbit Polyclonal Antibody
Antibodies
ATP6V1D is also named as ATP6M, VATD(V-type proton ATPase subunit D) and belongs to the V-ATPase D subunit family. ATP6V1D gene has been under strong negative selection during evolution and is highly conserved among mammals, flies, worms, yeast, plants, and bacteria. It is responsible for acidifying a variety of intracellular compartments in eukaryotic cells, thus providing most of the energy required for transport processes in the vacuolar system.

Western Blot: Human Brain Tissue, 1:500-1:5000; IHC: Human lung cancer Tissue, 1:20-1:200; FC: HeLa Cells, N/A

Type: Primary
Antigen: ATP6V1D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR