Anti-RPS26 Rabbit Polyclonal Antibody

Supplier: Proteintech
14909-1-AP-150UL
10093-956EA 402 USD
10093-956
Anti-RPS26 Rabbit Polyclonal Antibody
Antibodies
Diamond-Blackfan anemia (DBA) is a congenital red cell aplasia that is usually diagnosed during early infancy, and strikingly, in addition to defects in red cell maturation the disorder is associated with various physical anomalies in about 40% of patients. To date, heterozygous mutations in nine RP genes have been conclusively associated with DBA, and RPS26 is one of them. There are nine mutation found in RPS26 from DBA patients.

Western Blot: Mouse ovary Tissue, 1:500-1:5000; IF: MCF-7 Cells, 1:20-1:200; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: RPS26
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR