Anti-FADD Rabbit Polyclonal Antibody

Supplier: Proteintech
14906-1-AP-150UL
10086-758EA 402 USD
10086-758
Anti-FADD Rabbit Polyclonal Antibody
Antibodies
Western Blot: HT-1080 Cells, 1:200-1:1000; IHC: Human lung cancer Tissue, 1:20-1:200; IP:A549 Cells, 1:200-1:1000

Type: Primary
Antigen: FADD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR