Anti-GSTZ1 Rabbit Polyclonal Antibody

Supplier: Proteintech
14889-1-AP-150UL
10087-946EA 402 USD
10087-946
Anti-GSTZ1 Rabbit Polyclonal Antibody
Antibodies
Glutathione transferase zeta 1(GSTZ1) catalyzes the biotransformation of a range of α-haloacids, including dichloroacetic acid (DCA), and the penultimate step in the tyrosine degradation pathway. GSTZ1 is preferentially expressed in hepatocytes and renal proximal tubule cells where phenylalanine and tyrosine are catabolized. This protein has 3 isoforms produced by alternative splicing.

Western Blot: Mouse Liver Tissue, 1:500-1:5000

Type: Primary
Antigen: GSTZ1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR