Anti-C1orf116 Rabbit Polyclonal Antibody

Supplier: Proteintech
14888-1-AP-150UL
10083-802EA 402 USD
10083-802
Anti-C1orf116 Rabbit Polyclonal Antibody
Antibodies
Specifically androgen-regulated gene protein (SARG), encoded by C1orf116, is a 601-aa protein considered putative androgen-specific receptor. SARG mRNA is highly expressed in prostate and can be up-regulated by androgens, but not by glucocorticoids. Little is known about the function of SARG to date. Two isoforms of SARG are produced by alternative splicing encoding 64 kDa and 38 kDa proteins, and both isoforms can be recognized by this antibody 14888-1-AP.

Western Blot:PC-3 Cells, 1:200-1:2000

Type: Primary
Antigen: C1orf116
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR