Anti-NARS Rabbit Polyclonal Antibody

Supplier: Proteintech
14882-1-AP-150UL
10090-820EA 402 USD
10090-820
Anti-NARS Rabbit Polyclonal Antibody
Antibodies
Western Blot: HepG2 Cells, 1:1000-1:10000; IHC: Human Liver cancer, 1:20-1:200

Type: Primary
Antigen: NARS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR