Anti-COQ9 Rabbit Polyclonal Antibody

Supplier: Proteintech
14874-1-AP-150UL
10085-028EA 524.48 USD
10085-028
Anti-COQ9 Rabbit Polyclonal Antibody
Antibodies
COQ9, also named as C16orf49, HSPC326, and PSEC0129, is a 318 amino acid protein, which belongs to the COQ9 family. COQ9 localizes in the Mitochondrion and is involved in the biosynthesis of coenzyme Q COQ9 has abilities to improve left ventricular performance and to reduce myocardial infarct size and cardiomyocyte apoptosis.

Western Blot: Human Kidney Tissue, 1:500-1:5000; IHC: Human cervical cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; IP: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: COQ9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR