Anti-SDHA Rabbit Polyclonal Antibody

Supplier: Proteintech
14865-1-AP-150UL
10094-232EA 402 USD
10094-232
Anti-SDHA Rabbit Polyclonal Antibody
Antibodies
SDHA, also named as SDH2, SDHF and Fp, belongs to the FAD-dependent oxidoreductase 2 family and FRD/SDH subfamily. SDHA subunit of succinate dehydrogenase (SDH) that is involved in complex II of the mitochondrial electron transport chain and is responsible for transferring electrons from succinate to ubiquinone (coenzyme Q). It can be detected a band of 68 kDa in cutaneous melanoma. It is act as a tumor suppressor.

Western Blot: HEK-293 Cells, 1:200-1:2000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: SDHA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR