Anti-Chymotrypsin like protease Rabbit Polyclonal Antibody

Supplier: Proteintech
14856-1-AP-150UL
10085-328EA 402 USD
10085-328
Anti-Chymotrypsin like protease Rabbit Polyclonal Antibody
Antibodies
CTRL(Chymotrypsin-like protease) is also named as CTRL1 and belongs to the peptidase S1 family. The CTRL1 enzyme is active over a broad pH range. It expresses in pancreas and presence in intestinal fluid after stimulation of pancreatic secretion as a digestive enzyme. This gene encode a protein of 264 amino acid and the protein has a signal peptide of 18 amino acid and a propeptide of 15 amino acid.

Western Blot: Mouse pancreas Tissue, 1:1000-1:10000; IHC: Human pancreas Tissue, 1:20-1:200; IF:BxPC-3 Cells, 1:10-1:100

Type: Primary
Antigen: Chymotrypsin like protease
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR