Anti-CRISP3 Rabbit Polyclonal Antibody

Supplier: Proteintech
14847-1-AP-150UL
10082-056EA 402 USD
10082-056
Anti-CRISP3 Rabbit Polyclonal Antibody
Antibodies
CRISP3, also known as SGP28, is a glycoprotein that belongs to a family of cysteine-rich secretory proteins (CRISPs). CRISP3 was originally discovered in human neutrophils, but transcripts are widely distributed in exocrine glands (salivary glands, pancreas, and prostate) and also found at lower levels in epididymis, ovary, thymus, and colon. CRISP3 is a potential biomarker for prostate cancer. This antibody can recgnize both CRISP2 and CRISP3.

Western Blot: Human Placenta Tissue, 1:500-1:5000; IHC: Human pancreas Tissue, 1:20-1:200

Type: Primary
Antigen: CRISP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR