Anti-FABP7 Rabbit Polyclonal Antibody

Supplier: Proteintech
14836-1-AP-150UL
10086-408EA 402 USD
10086-408
Anti-FABP7 Rabbit Polyclonal Antibody
Antibodies
FABP7 (Fatty Acid Binding Protein-7), also known as B-FABP (Brain Fatty Acid-Binding Protein), is a member of FABP family which are intracellular proteins involved in lipid metabolism. Normally FABP7 is mainly expressed in brain and other neural tissues and is important in early nervous system development. In addition, FABP7 is considered a marker for some brain tumors and may have a role in tumorigenesis in the peripheral nervous system and in several extra nervous system neoplasms such as breast cancer, renal cancer, and melanoma. This antibody specially recognizes FABP7 and not cross-react with other FABPs.

Western Blot: Human Brain Tissue, 1:1000-1:10000; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: FABP7-Specific
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR