Anti-GLYR1 Rabbit Polyclonal Antibody

Supplier: Proteintech
14833-1-AP-150UL
10087-648EA 402 USD
10087-648
Anti-GLYR1 Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:1000-1:10000; IHC: Human Colon Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: GLYR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR