Anti-RPS6 Rabbit Polyclonal Antibody

Supplier: Proteintech
14823-1-AP-150UL
10093-978EA 402 USD
10093-978
Anti-RPS6 Rabbit Polyclonal Antibody
Antibodies
Ribosomal protein S6 (RPS6),Phosphoprotein NP33.It may play an important role in controlling cell growth and proliferation through the selective translation of particular classes of mRNA.Ribosomal protein S6 is the major substrate of protein kinases in eukaryote ribosomes. The phosphorylation is stimulated by growth factors, tumor promoting agents, and mitogens. It is dephosphorylated at growth arrest. Phosphorylated at Ser-235 and Ser-236 by RPS6KA1 and RPS6KA3; phosphorylation at these sites facilitates the assembly of the preinitiation complex.It's molecular weight is about 29kd.

Western Blot:K-562 Cells, 1:200-1:2000

Type: Primary
Antigen: RPS6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR