Anti-DEXI Rabbit Polyclonal Antibody

Supplier: Proteintech
14811-1-AP-150UL
10085-778EA 402 USD
10085-778
Anti-DEXI Rabbit Polyclonal Antibody
Antibodies
IHC: Human lung cancer, 1:20-1:200

Type: Primary
Antigen: DEXI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR