Anti-CCDC28A Rabbit Polyclonal Antibody

Supplier: Proteintech
14807-1-AP-150UL
10084-180EA 402 USD
10084-180
Anti-CCDC28A Rabbit Polyclonal Antibody
Antibodies
Western Blot:A375 Cells, 1:500-1:5000; IF: MCF-7, 1:20-1:200

Type: Primary
Antigen: CCDC28A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR