Anti-NLN Rabbit Polyclonal Antibody

Supplier: Proteintech
14763-1-AP-150UL
10091-142EA 402 USD
10091-142
Anti-NLN Rabbit Polyclonal Antibody
Antibodies
NLN(Neurolysin, mitochondrial) is also named as AGTBP, KIAA1226 and belongs to the peptidase M3 family. It is an enzyme involved in the metabolic inactivation of bioactive peptides and functions as a monomer of molecular mass 78 kDa. It also can be dected the molecular weight between 70 kDa and 85 kDa according to the reports . The full length protein has a transit peptide with 37 amino acids.

Western Blot: Mouse ovary Tissue, 1:200-1:1000; IHC: Human ovary tumor Tissue, 1:20-1:200

Type: Primary
Antigen: NLN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR