Anti-CDK5RAP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
14740-1-AP-150UL
10084-526EA 402 USD
10084-526
Anti-CDK5RAP1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Heart Tissue, 1:500-1:5000; IHC: Human pancreas Tissue, 1:20-1:200

Type: Primary
Antigen: CDK5RAP1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR