Anti-TMEM98 Rabbit Polyclonal Antibody

Supplier: Proteintech
14731-1-AP-150UL
10095-702EA 402 USD
10095-702
Anti-TMEM98 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse E12.5 Embryos, N/A

Type: Primary
Antigen: TMEM98
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR