Anti-SPAG5 Rabbit Polyclonal Antibody

Supplier: Proteintech
14726-1-AP-150UL
10095-046EA 402 USD
10095-046
Anti-SPAG5 Rabbit Polyclonal Antibody
Antibodies
SPAG5, also named as MAP126, Astrin and Deepest, is necessary for normal spindle formation during mitosis. It is highly expressed in testis. SPAG5 has different localization in somatic versus male germ cells suggesting the possibility of different function. This antibody is specific to SPAG5.

Western Blot: HeLa Cells, 1:1000-1:10000; IHC: Human Testis Tissue, 1:20-1:200; IF:A431 Cells, 1:20-1:200; IP: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: SPAG5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR