Anti-AHSA1 Rabbit Polyclonal Antibody

Supplier: Proteintech
14725-1-AP-150UL
10082-630EA 537.7 USD
10082-630
Anti-AHSA1 Rabbit Polyclonal Antibody
Antibodies
Western Blot:A549 Cells, 1:200-1:2000; IHC: Human gliomas Tissue, 1:20-1:200; IF:A431 Cells, 1:20-1:200

Type: Primary
Antigen: AHSA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR