Anti-CNBP Rabbit Polyclonal Antibody

Supplier: Proteintech
14717-1-AP-150UL
10084-914EA 537.7 USD
10084-914
Anti-CNBP Rabbit Polyclonal Antibody
Antibodies
Single stranded DNA-binding protein, with specificity to the sterol regulatory element (SRE). Involved in sterol-mediated repression.

IHC: Human ovary Tissue, 1:20-1:200; Western Blot: Mouse Testis Tissue, 1:200-1:1000

Type: Primary
Antigen: CNBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR