Anti-MAP4K3 Rabbit Polyclonal Antibody

Supplier: Proteintech
14702-1-AP-150UL
10089-946EA 402 USD
10089-946
Anti-MAP4K3 Rabbit Polyclonal Antibody
Antibodies
MAP4K3(Mitogen-activated protein kinase kinase kinase kinase 3) has kinase activity and that it activates JNK but not MAPK1, MAPK3 or MAPK14 through the activation of MAP3K1 and MAP2K4. Endogenous MAP4K3 kinase activity could be stimulated by exposure to ultraviolet radiation or to tumor necrosis factor.

Western Blot: Human Skeletal Muscle Tissue, 1:1000-1:10000; IHC: Human Skeletal Muscle Tissue, 1:20-1:200

Type: Primary
Antigen: MAP4K3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR