Anti-Collagen Type I Rabbit Polyclonal Antibody

Supplier: Proteintech
14695-1-AP-150UL
10084-982EA 402 USD
10084-982
Anti-Collagen Type I Rabbit Polyclonal Antibody
Antibodies
Type I collagen, the major structural component of connective tissues such as skin, tendon and bone, is the most abundant and widely expressed collagen in humans . Type I collagen is a heterotrimer comprising one alpha 2(I) and two alpha 1(I) chains which are encoded by the unlinked loci COL1A2 and COL1A1 respectively. Mutations in COL1A2 gene are associated with osteogenesis imperfecta, Ehlers-Danlos syndrome, idiopathic osteoporosis, and atypical Marfan syndrome.

Western Blot: Human Kidney Tissue, 1:200-1:2000

Type: Primary
Antigen: Collagen Type I
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR