Anti-RXRB Rabbit Polyclonal Antibody

Supplier: Proteintech
14684-1-AP-150UL
10093-740EA 402 USD
10093-740
Anti-RXRB Rabbit Polyclonal Antibody
Antibodies
Western Blot:NIH/3T3 Cells, 1:200-1:2000

Type: Primary
Antigen: RXRB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR