Anti-MTA3 Rabbit Polyclonal Antibody

Supplier: Proteintech
14682-1-AP-150UL
10090-530EA 524.48 USD
10090-530
Anti-MTA3 Rabbit Polyclonal Antibody
Antibodies
Metastasis-associated protein 3 (MTA3) is a cell-type specific component of the Mi-2-NURD transcriptional co-repressor complex. It has a role in maintenance of the normal epithelial architecture through the repression of SNAI1 transcription in a histone deacetylase-dependent manner, and thus the regulation of E-cadherin levels.

Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human Brain, 1:20-1:200; IF: HepG2, 1:20-1:200

Type: Primary
Antigen: MTA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR