Anti-ASNS Rabbit Polyclonal Antibody

Supplier: Proteintech
14681-1-AP-150UL
10083-202EA 537.7 USD
10083-202
Anti-ASNS Rabbit Polyclonal Antibody
Antibodies
Asparagine synthetase [glutamine-hydrolyzing](ASNS) is also named as TS11. The asparagine synthetase gene, encoding the enzyme that catalyzes the synthesis of asparagine and glutamate using glutamine and aspartate, is a gene for which transcription is highly regulated by the nutritional status of the cell. It encodes a protein of about 561 amino acids with the predicted molecular mass of 64 kDa.

Western Blot:Jurkat Cells, 1:500-1:5000

Type: Primary
Antigen: ASNS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR