Anti-PHLDA2 Rabbit Polyclonal Antibody

Supplier: Proteintech
14661-1-AP-150UL
10092-208EA 453 USD
10092-208
Anti-PHLDA2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Placenta Tissue, 1:500-1:5000

Type: Primary
Antigen: PHLDA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR