Anti-RDH10 Rabbit Polyclonal Antibody

Supplier: Proteintech
14644-1-AP-150UL
10093-474EA 537.7 USD
10093-474
Anti-RDH10 Rabbit Polyclonal Antibody
Antibodies
RDH10(Retinol dehydrogenase 10) is a strictly NAD+-dependent enzyme with multisubstrate specificity that recognizes cis-retinols as well as all-trans-retinol as substrates. RDH10-mediated oxidation of retinol plays as important a role in the control and regulation of RA production during embryogenesis as does the subsequent RALDH-mediated reaction.

Western Blot: Human Kidney Tissue, 1:500-1:5000; IHC: Human lung cancer Tissue, 1:20-1:200; IP: Mouse Liver Tissue, 1:200-1:2000

Type: Primary
Antigen: RDH10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR