Anti-DDX6 Rabbit Polyclonal Antibody

Supplier: Proteintech
14632-1-AP-150UL
10091-694EA 402 USD
10091-694
Anti-DDX6 Rabbit Polyclonal Antibody
Antibodies
P54, also named as DDX6, HLR2, and RCK, is a 483 amino acid protein, which contains one helicase C-terminal domain, one helicase ATP-binding domain, and belongs to the DEAD box helicase family. DDX6/DHH1 subfamily. P54 is abundantly expressed in most tissues. In the process of mRNA degradation, P54 may play a role in mRNA decapping. P54 is involved in the replication of hepatitis C virus genomes in hepatocytes and in tumourigenesis of hepatocellular carcinomas.

Western Blot:K-562 Cells, 1:500-1:5000; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: DDX6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR