Anti-DUSP15 Rabbit Polyclonal Antibody

Supplier: Proteintech
14629-1-AP-150UL
10086-176EA 402 USD
10086-176
Anti-DUSP15 Rabbit Polyclonal Antibody
Antibodies
Western Blot:A375 Cells, 1:200-1:1000

Type: Primary
Antigen: DUSP15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR