Anti-MYO18A Rabbit Polyclonal Antibody

Supplier: Proteintech
14611-1-AP-150UL
10090-740EA 402 USD
10090-740
Anti-MYO18A Rabbit Polyclonal Antibody
Antibodies
MYO18A is an unconventional myosin that may play a role in the stabilization and organization of the actin cytoskeleton. Recently MYO18A has been identified as the receptor of SP and is responsible for clearance of SP-A-opsonized bacteria in in vivo infection. Two different isoforms of mouse Myo18A have been identified: Myo18A-alpha and Myo18A-beta as 230- and 190-kD proteins, respectively.Myo18a-alpha colocalizes with the ER-Golgi complex and with membrane ruffles and filopodia, while Myo18a-beta localizes diffusely to the cytoplasm.

Western Blot:Jurkat Cells, 1:1000-1:10000; IP: HeLa Cells, 1:200-1:1000

Type: Primary
Antigen: MYO18A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR