Anti-DNM2 Rabbit Polyclonal Antibody

Supplier: Proteintech
14605-1-AP-150UL
10086-042EA 459.7 USD
10086-042
Anti-DNM2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HL-60 Cells, 1:200-1:2000

Type: Primary
Antigen: DNM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR