Anti-BRN2 Rabbit Polyclonal Antibody

Supplier: Proteintech
14596-1-AP-150UL
10083-690EA 402 USD
10083-690
Anti-BRN2 Rabbit Polyclonal Antibody
Antibodies
POU3F2 belongs to a large family of transcription factors that bind to the octameric DNA sequence ATGCAAAT. It shares a highly homologous region that referred to as the POU domain. POU3F2 is a Class III POU genes that expressed predominantly in the central nervous system (CNS). It is likely that CNS-specific transcription factors such as these play an important role in mammalian neurogenesis by regulating their diverse patterns of gene expression. POU3F2 is required for maintaining neural cell differetiation and has a role in the production and positioning of neocortical neurons.

IHC: Human gliomas Tissue, 1:20-1:200; Western Blot:Jurkat Cells, 1:500-1:5000

Type: Primary
Antigen: BRN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR