Anti-FBXO2 Rabbit Polyclonal Antibody

Supplier: Proteintech
14590-1-AP-150UL
10086-906EA 402 USD
10086-906
Anti-FBXO2 Rabbit Polyclonal Antibody
Antibodies
IHC: Human Brain Tissue, 1:20-1:200; Western Blot:SH-SY5Y Cells, 1:200-1:1000

Type: Primary
Antigen: FBXO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR