Anti-IKIP Rabbit Polyclonal Antibody

Supplier: Proteintech
14589-1-AP-150UL
10088-690EA 402 USD
10088-690
Anti-IKIP Rabbit Polyclonal Antibody
Antibodies
IKIP(Inhibitor of nuclear factor kappa-B kinase-interacting protein) is also named as IKBIP and acts as a novel p53 target gene with proapoptotic function. A variety of experiments looking at endogenous IKIP1 levels showed that IKIP1 expression is dependent on p53. Transfection of IKIP into endothelial cells promotes cell death, as measured by TUNEL staining. EST analysis and RT-PCR demonstrated that 3 IKIP transcripts are generated; IKIP1 encodes a 377-amino acid protein, IKIP2 a 350-amino acid protein, and IKIP3 2 potential ORFs encoding 70- and 245-amino acid proteins.

Western Blot: Mouse Skeletal Muscle Tissue, 1:200-1:2000

Type: Primary
Antigen: IKIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR