Anti-EGFLAM Rabbit Polyclonal Antibody

Supplier: Proteintech
14578-1-AP-150UL
10086-308EA 524.48 USD
10086-308
Anti-EGFLAM Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Placenta Tissue, 1:500-1:5000

Type: Primary
Antigen: EGFLAM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR